Diethyl-3,5-di-tert-butyl-4-hydroxybenzylphosphonate ≥98.0% (by HPLC)
Supplier: TCI America
D2967-1G
D2967-5G
TCD2967-1GEA
121.66
CAD
TCD2967-1G
TCD2967-5G
Diethyl-3,5-di-tert-butyl-4-hydroxybenzylphosphonate ≥98.0% (by HPLC)
Diethyl-3,5-di-tert-butyl-4-hydroxybenzylphosphonate
CAS Number: 976-56-7
MDL Number: MFCD00227871
Molecular Formula: C19H33O4P
Molecular Weight: 356.44
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 122
Other Product Names / Synonyms: 3,5-Di-tert-butyl-4-hydroxybenzylphosphonic Acid Diethyl Ester
Formula:
C₁₉H₃₃O₄P Melting Pt: 122 °C |
MDL Number:
MFCD00227871 CAS Number: 976-56-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...