Di-n-octyl sulfoxide ≥98.0% (by HPLC)

Supplier: TCI America

D2965-5G D2965-1G
TCD2965-5GEA 289.3 CAD
TCD2965-5G TCD2965-1G
Di-n-octyl sulfoxide ≥98.0% (by HPLC)
Di-n-octyl sulfoxide

CAS Number: 1986-89-6
MDL Number: MFCD00039514
Molecular Formula: C16H34OS
Molecular Weight: 274.51
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 71


Other Product Names / Synonyms: n-Octyl Sulfoxide

Formula: C₁₆H₃₄OS
Melting Pt: 71 °C
MDL Number: MFCD00039514
CAS Number: 1986-89-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a736b3b0beec3d70ae7f76927b2f68215c8bb18&vid=3a736b3b0beec3d70ae7f76927b2f68215c8bb18&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR