N,N'-Dimethoxy-N,N'-dimethyloxamide ≥98.0%
Supplier: TCI America
Synonyms:
N1,N2-Dimethoxy-N1,N2-dimethyloxalamide, N,N'-Oxalylbis[methoxy(methyl)amine]
D2961-25G
D2961-5G
TCD2961-25GEA
643.06
CAD
TCD2961-25G
TCD2961-5G
N,N'-Dimethoxy-N,N'-dimethyloxamide ≥98.0%
N,N'-Dimethoxy-N,N'-dimethyloxamide
CAS Number: 106675-70-1
MDL Number: MFCD02326951
Molecular Formula: C6H12N2O4
Molecular Weight: 176.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 93
Other Product Names / Synonyms: N,N'-Oxalylbis[methoxy(methyl)amine]
Formula:
C₆H₁₂N₂O₄ MW: 176.17 g/mol Melting Pt: 93 °C |
MDL Number:
MFCD02326951 CAS Number: 106675-70-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...