6-Amidino-2-naphthol Methanesulfonate ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 6-Amidino-2-naphthol Methanesulphonic Acid, 6-Hydroxynaphthalene-2-carboximidamide Methanesulphonate, 6-Amidino-2-naphthol Methanesulfonic acid, 6-Hydroxynaphthalene-2-carboximidamide Methanesulfonate, 6-Hydroxy-2-naphthimidamide methanesulphonate salt, 6-Amidino-2-naphthol Methanesulphonate, 6-Hydroxy-2-naphthimidamide methanesulfonate salt, 6-Carbamimidoyl-2-naphthol Methanesulphonate, 6-Amidino-2-naphthol Methanesulfonic Acid, 6-Carbamimidoyl-2-naphthol Methanesulfonate, 6-Amidino-2-naphthol Methanesulfonate

A1193-1G A1193-5G
TCA1193-001GEA 243.32 CAD
TCA1193-001G TCA1193-5G
6-Amidino-2-naphthol Methanesulfonate ≥99.0% (by HPLC, titration analysis)
6-Amidino-2-naphthol Methanesulfonate

CAS Number: 82957-06-0
MDL Number: MFCD00143493
Molecular Formula: C11H10N2O
Molecular Weight: 282.31
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 6-Amidino-2-naphthol Methanesulfonic Acid, 6-Hydroxynaphthalene-2-carboximidamide Methanesulfonate, 6-Carbamimidoyl-2-naphthol Methanesulfonate

Formula: C₁₁H₁₀N₂O
MW: 282.31 g/mol
MDL Number: MFCD00143493
CAS Number: 82957-06-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ec6eb900842c01c69ac98ed333a69d3bb2f2f7b&vid=1ec6eb900842c01c69ac98ed333a69d3bb2f2f7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR