(±)-2-Dodecanol ≥98.0%

Supplier: TCI America

D2960-5G D2960-25G
TCD2960-5GEA 262.28 CAD
TCD2960-5G TCD2960-25G
(±)-2-Dodecanol ≥98.0%
(±)-2-Dodecanol

CAS Number: 10203-28-8
MDL Number: MFCD00004551
Molecular Formula: C12H26O
Molecular Weight: 186.34
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 129
Melting point (°C): 19
Flash Point (°C): 110
Specific Gravity (20/20): 0.83

Formula: C₁₂H₂₆O
Boiling Pt: 129 °C
Melting Pt: 19 °C
Density: 0.83 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00004551
CAS Number: 10203-28-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3dd12dc6c08f7e8654857b65971a325b66ff073&vid=2905d0d8ac59ba6a92b206b6b1b26ba2a9c12234&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR