N,N-Diethyl-2,3-dimethylbenzamide ≥98.0%

Supplier: TCI America

D2956-25G D2956-5G
TCD2956-25GEA 279.66 CAD
TCD2956-25G TCD2956-5G
N,N-Diethyl-2,3-dimethylbenzamide ≥98.0%
N,N-Diethyl-2,3-dimethylbenzamide

CAS Number: 57806-76-5
MDL Number: MFCD06797096
Molecular Formula: C13H19NO
Molecular Weight: 205.30
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: N,N-Diethyl-o-xylene-3-carboxamide

Formula: C₁₃H₁₉NO
Density: 1.00 (20 °C)
MDL Number: MFCD06797096
CAS Number: 57806-76-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18c451103bf7108404b7ce1c48b769c87d42a081&vid=18c451103bf7108404b7ce1c48b769c87d42a081&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR