Divinyl adipate ≥99.0% stabilized

Supplier: TCI America

A1188-25G A1188-100G
TCA1188-025GEA 96.22 CAD
TCA1188-025G TCA1188-500G TCA1188-100G
Divinyl adipate ≥99.0% stabilized
Divinyl adipate

(stabilized with MEHQ)
CAS Number: 4074-90-2
MDL Number: MFCD00059202
Molecular Formula: C10H14O4
Molecular Weight: 198.22
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Flash Point (°C): 116
Freezing point (°C): 28


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Adipic Acid Divinyl Ester

Formula: C₁₀H₁₄O₄
Flash Pt: 116 °C
MDL Number: MFCD00059202
CAS Number: 4074-90-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4fd916d43ef07094997816558e3c529a3fe7009&vid=d4fd916d43ef07094997816558e3c529a3fe7009&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR