(2,3-Dichlorophenyl)acetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2,3-Dichlorophenylacetic acid

D2931-25G D2931-5G
TCD2931-25GEA 431.34 CAD
TCD2931-25G TCD2931-5G
(2,3-Dichlorophenyl)acetic acid ≥98.0% (by GC, titration analysis)
(2,3-Dichlorophenyl)acetic acid

CAS Number: 10236-60-9
MDL Number: MFCD01861393
Molecular Formula: C8H6Cl2O2
Molecular Weight: 205.03
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 134

Formula: (C₆H₃Cl₂)CH₂COOH
MW: 205.04 g/mol
Melting Pt: 134 °C
MDL Number: MFCD01861393
CAS Number: 10236-60-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dabf41e5c219ea67ad438c86f60350d52afff36&vid=5dabf41e5c219ea67ad438c86f60350d52afff36&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR