2,6-Dichlorophenethylamine ≥98.0%
Supplier: TCI America
Synonyms:
2-(2-Aminoethyl)-1,3-dichlorobenzene, 1-Amino-2-(2,6-dichlorophenyl)ethane, 2-(2,6-Dichlorophenyl)ethylamine, 2,6-Dichlorophenethylamine
CAS Number: 14573-23-0
MDL Number: MFCD00060612
Molecular Formula: C8H9Cl2N
Molecular Weight: 190.07
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 129
Flash Point (°C): 110
Specific Gravity (20/20): 1.28
Other Product Names / Synonyms: 1-Amino-2-(2,6-dichlorophenyl)ethane, 2-(2-Aminoethyl)-1,3-dichlorobenzene, 2,6-Dichlorophenethylamine
Formula:
C₈H₉Cl₂N MW: 190.07 g/mol Boiling Pt: 129 °C Density: 1.28 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00060612 CAS Number: 14573-23-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...