(2,4-Dichlorobenzyl)triphenylphosphonium chloride ≥98.0%

Supplier: TCI America

Synonyms: Triphenyl(2,4-dichlorobenzyl)phosphonium Chloride

D2907-25G D2907-5G
TCD2907-25GEA 185.18 CAD
TCD2907-25G 76219-012
(2,4-Dichlorobenzyl)triphenylphosphonium chloride ≥98.0%
(2,4-Dichlorobenzyl)triphenylphosphonium chloride

CAS Number: 2492-23-1
MDL Number: MFCD00031641
Molecular Formula: C25H20Cl3P
Molecular Weight: 457.76
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 249


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: Triphenyl(2,4-dichlorobenzyl)phosphonium Chloride

Formula: C₂₅H₂₀Cl₃P
MW: 457.76 g/mol
Melting Pt: 249 °C
MDL Number: MFCD00031641
CAS Number: 2492-23-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55e88dd4ba8e7b77d7efe7af758d7d58911f16ad&vid=55e88dd4ba8e7b77d7efe7af758d7d58911f16ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR