Dodecaethylene glycol monomethyl ether ≥90.0%

Supplier: TCI America

D2904-100MG D2904-1G
TCD2904-100MGEA 323.9 CAD
TCD2904-100MG TCD2904-1G
Dodecaethylene glycol monomethyl ether ≥90.0%
Dodecaethylene glycol monomethyl ether

MDL Number: MFCD06797088
Molecular Formula: C25H52O13
Molecular Weight: 560.68
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid


Other Product Names / Synonyms: mPEG12-Alcohol

Formula: C₂₅H₅₂O₁₃
MDL Number: MFCD06797088
CAS Number: 5702-16-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3227d7dffc905fd40d912fd716dcd3ef6d095bb9&vid=3227d7dffc905fd40d912fd716dcd3ef6d095bb9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR