Didodecyl sulfoxide ≥98.0%

Supplier: TCI America

Synonyms: Dodecyl sulphoxide, Dodecyl sulfoxide

D2885-25G D2885-5G
TCD2885-25GEA 885.91 CAD
TCD2885-25G TCD2885-5G
Didodecyl sulfoxide ≥98.0%
Didodecyl sulphoxide

CAS Number: 2168-96-9
MDL Number: MFCD00039865
Molecular Formula: C24H50OS
Molecular Weight: 386.72
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 91


Other Product Names / Synonyms: Dodecyl Sulfoxide

Formula: C₂₄H₅₀OS
Melting Pt: 91 °C
MDL Number: MFCD00039865
CAS Number: 2168-96-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78b93c4b28bae4d3a1c8ef85440200af01f8c3e7&vid=78b93c4b28bae4d3a1c8ef85440200af01f8c3e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR