Methyl-2,5-dihydrothiophene-3-carboxylate ≥95.0%
Supplier: TCI America
Synonyms:
Methyl 2,5-dihydrothiophene-3-carboxylate, 2,5-Dihydrothiophene-3-carboxylic Acid Methyl Ester
D2882-1G
D2882-5G
TCD2882-1GEA
60.04
CAD
TCD2882-1G
TCD2882-5G
Methyl-2,5-dihydrothiophene-3-carboxylate ≥95.0%
Methyl-2,5-dihydrothiophene-3-carboxylate
CAS Number: 67488-46-4
MDL Number: MFCD04038417
Molecular Formula: C6H8O2S
Molecular Weight: 144.19
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 36
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 2,5-Dihydrothiophene-3-carboxylic Acid Methyl Ester
Formula:
C₆H₈O₂S MW: 144.19 g/mol Melting Pt: 36 °C |
MDL Number:
MFCD04038417 CAS Number: 67488-46-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...