Diamantane ≥98.0%
Supplier: TCI America
Synonyms:
Pentacyclo[7.3.1.1(4,12).0(2,7).0(6,11)]tetradecane, Congressane
D2876-1G
D2876-5G
TCD2876-100MGEA
47.19
CAD
TCD2876-100MG
TCD2876-1G
TCD2876-5G
Diamantane ≥98.0%
Diamantane
CAS Number: 2292-79-7
MDL Number: MFCD00171301
Molecular Formula: C14H20
Molecular Weight: 188.31
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 245
Other Product Names / Synonyms: Congressane, Pentacyclo[7.3.1.1(4,12).0(2,7).0(6,11)]tetradecane
Formula:
C₁₄H₂₀ MW: 188.31 g/mol Melting Pt: 245 °C |
MDL Number:
MFCD00171301 CAS Number: 2292-79-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...