5-Amino-2-methoxybenzenesulfonic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A1177-25G A1177-5G
TCA1177-025GEA 358.66 CAD
TCA1177-025G TCA1177-5G
5-Amino-2-methoxybenzenesulfonic acid ≥98.0% (by HPLC, titration analysis)
5-Amino-2-methoxybenzenesulphonic acid

CAS Number: 6470-17-3
MDL Number: MFCD00035760
Molecular Formula: C7H9NO4S
Molecular Weight: 203.21
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 4-Aminoanisole-2-sulfonic Acid, 3-Amino-6-methoxybenzenesulfonic Acid, 4-Methoxyaniline-3-sulfonic Acid

Formula: C₇H₉NO₄S
MDL Number: MFCD00035760
CAS Number: 6470-17-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c390f3cdab3d18c4a6152f54f2a749c9324efa0&vid=8c390f3cdab3d18c4a6152f54f2a749c9324efa0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR