2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥98.0%

Supplier: TCI America

Synonyms: 3,5-Dimethyl-4-hydroxyphenylboronic acid pinacol ester, 3,5-Dimethy-4-hydroxyphenylboronic acid pinacol ester

D2853-5G D2853-1G
TCD2853-5GEA 557.74 CAD
TCD2853-5G TCD2853-1G
2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥98.0%
3,5-Dimethyl-4-hydroxyphenylboronic acid pinacol ester

CAS Number: 269410-25-5
MDL Number: MFCD02093724
Molecular Formula: C14H21BO3
Molecular Weight: 248.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 108


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 2-(4-Hydroxy-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,6-xylenol, 4-Hydroxy-3,5-dimethylphenylboronic Acid Pinacol Ester

Formula: C₁₄H₂₁BO₃
MW: 248.13 g/mol
Melting Pt: 108 °C
MDL Number: MFCD02093724
CAS Number: 269410-25-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67243d2066238d6cf895bee085ef5efec065c054&vid=67243d2066238d6cf895bee085ef5efec065c054&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR