3,7-Dithia-1,9-nonanedithiol ≥97.0% (by GC, titration analysis)

Supplier: TCI America

D2852-5G D2852-1G
TCD2852-5GEA 1085.46 CAD
TCD2852-5G TCD2852-1G
3,7-Dithia-1,9-nonanedithiol ≥97.0% (by GC, titration analysis)
3,7-Dithia-1,9-nonanedithiol

CAS Number: 25676-62-4
MDL Number: MFCD00506918
Molecular Formula: C7H16S4
Molecular Weight: 228.45
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 1.18


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,3-Bis(2-mercaptoethylthio)propane, 2,2'-(Trimethylenedithio)diethanethiol

Formula: C₇H₁₆S₄
Density: 1.18 (20 °C)
MDL Number: MFCD00506918
CAS Number: 25676-62-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18bddf164a0f87df116ae429b54ad436594ff9ec&vid=18bddf164a0f87df116ae429b54ad436594ff9ec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR