2,4-Dimethylpyrrole ≥98.0%
Supplier: TCI America
D2848-1G
D2848-5G
D2848-25G
TCD2848-1GEA
61.15
CAD
TCD2848-1G
TCD2848-5G
76219-010
2,4-Dimethylpyrrole ≥98.0%
2,4-Dimethylpyrrole
CAS Number: 625-82-1
MDL Number: MFCD00192088
Molecular Formula: C6H9N
Molecular Weight: 95.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 0.92
Storage Temperature: 0-10°C
Formula:
C₆H₉N Density: 0.92 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00192088 CAS Number: 625-82-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...