2,2-Di-n-octyl-1,3-propanediol ≥95.0%

Supplier: TCI America

Synonyms: 9,9-Bis(hydroxymethyl)heptadecane, 1,3-Dihydroxy-2,2-di-n-octylpropane

D2842-1G D2842-5G
TCD2842-1GEA 243 CAD
TCD2842-1G TCD2842-5G
2,2-Di-n-octyl-1,3-propanediol ≥95.0%
2,2-Di-n-octyl-1,3-propanediol

CAS Number: 106868-09-1
MDL Number: MFCD03844790
Molecular Formula: C19H40O2
Molecular Weight: 300.53
Purity/Analysis Method: >95.0% (GC)
Form: Crystal


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 9,9-Bis(hydroxymethyl)heptadecane, 1,3-Dihydroxy-2,2-di-n-octylpropane

Formula: C₁₉H₄₀O₂
MDL Number: MFCD03844790
CAS Number: 106868-09-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47e9ddb3e2fdb36b447512f6a5a3cf4ef8909b7e&vid=47e9ddb3e2fdb36b447512f6a5a3cf4ef8909b7e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR