(S)-2,2'-Dimethyl-1,1'-binaphthyl ≥98.0% (by HPLC)
Supplier: TCI America
D2838-1G
D2838-5G
TCD2838-1GEA
411.59
CAD
TCD2838-1G
TCD2838-5G
(S)-2,2'-Dimethyl-1,1'-binaphthyl ≥98.0% (by HPLC)
(S)-2,2'-Dimethyl-1,1'-binaphthyl
CAS Number: 32587-64-7
MDL Number: MFCD00048075
Molecular Formula: C22H18
Molecular Weight: 282.39
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Flash Point (°C): 110
Specific rotation [a]20/D: 17.5 deg (C=0.5, EtOH(95vol%))
Formula:
C₂₂H₁₈ Flash Pt: 110 °C |
MDL Number:
MFCD00048075 CAS Number: 32587-64-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...