(R)-2,2'-Dimethyl-1,1'-binaphthyl ≥98.0% (by HPLC)
Supplier: TCI America
D2837-1G
D2837-5G
TCD2837-1GEA
355.34
CAD
TCD2837-1G
TCD2837-5G
(R)-2,2'-Dimethyl-1,1'-binaphthyl ≥98.0% (by HPLC)
(R)-2,2'-Dimethyl-1,1'-binaphthyl
CAS Number: 19634-89-0
MDL Number: MFCD00048075
Molecular Formula: C22H18
Molecular Weight: 282.39
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 84
Flash Point (°C): 110
Specific rotation [a]20/D: -17.5 deg (C=0.5, EtOH(95vol%))
Formula:
C₂₂H₁₈ Melting Pt: 84 °C Flash Pt: 110 °C |
MDL Number:
MFCD00048075 CAS Number: 19634-89-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...