3',5'-Dibromo-4'-hydroxyacetophenone ≥97.0%

Supplier: TCI America

Synonyms: 1-(3,5-Dibromo-4-hydroxyphenyl)ethanone

D2824-25G D2824-5G
TCD2824-25GEA 480.32 CAD
TCD2824-25G TCD2824-5G
3',5'-Dibromo-4'-hydroxyacetophenone ≥97.0%
3',5'-Dibromo-4'-hydroxyacetophenone

CAS Number: 2887-72-1
MDL Number: MFCD00075779
Molecular Formula: C8H6Br2O2
Molecular Weight: 293.94
Purity/Analysis Method: >97.0% (GC)
Form: Crystal

Formula: C₈H₆Br₂O₂
MW: 293.9400 g/mol
MDL Number: MFCD00075779
CAS Number: 2887-72-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5b71423d43480472efaf072d77df7376be089fd&vid=c5b71423d43480472efaf072d77df7376be089fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR