(R,R)-2,2'-Isopropylidenebis(4-phenyl-2-oxazoline) ≥96.0%
Supplier: TCI America
Synonyms:
(R,R)-2,2-Bis(4-phenyl-2-oxazolin-2-yl)propane, (R,R)-2,2-Isopropylidenebis(4-phenyl-2-oxazoline), (R,R)-2,2'-(Dimethylmethylene)bis(4-phenyl-2-oxazoline)
CAS Number: 150529-93-4
MDL Number: MFCD00192244
Molecular Formula: C21H22N2O2
Molecular Weight: 334.42
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Specific rotation [a]20/D: 160 deg (C=1, EtOH)
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands
Other Product Names / Synonyms: (R,R)-2,2-Bis(4-phenyl-2-oxazolin-2-yl)propane, (R,R)-2,2'-(Dimethylmethylene)bis(4-phenyl-2-oxazoline)
Formula:
C₂₁H₂₂N₂O₂ MW: 334.42 g/mol |
MDL Number:
MFCD00192244 CAS Number: 150529-93-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...