2,6-Quinolinediol ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D2822-1G D2822-5G
TCD2822-1GEA 159.58 CAD
TCD2822-1G TCD2822-5G
2,6-Quinolinediol ≥98.0% (by HPLC, titration analysis)
2,6-Quinolinediol

CAS Number: 19315-93-6
MDL Number: MFCD00239446
Molecular Formula: C9H7NO2
Molecular Weight: 161.16
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 6-Hydroxy-2(1H)-quinolinone, 2,6-Quinolinediol

Formula: C₉H₇NO₂
MDL Number: MFCD00239446
CAS Number: 19315-93-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19a55bd8860658b614e4765629c308f411639f28&vid=19a55bd8860658b614e4765629c308f411639f28&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR