(S)-(-)-3,3'-Dibromo-1,1'-bi-2-naphthol ≥97.0% (by GC, titration analysis)
Supplier: TCI America
D2811-1G
D2811-5G
TCD2811-1GEA
421.54
CAD
TCD2811-1G
76219-008
(S)-(-)-3,3'-Dibromo-1,1'-bi-2-naphthol ≥97.0% (by GC, titration analysis)
(S)-(-)-3,3'-Dibromo-1,1'-bi-2-naphthol
CAS Number: 119707-74-3
MDL Number: MFCD03093629
Molecular Formula: C20H12Br2O2
Molecular Weight: 444.12
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Specific rotation [a]20/D: -90 deg (C=1, MeOH)
Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis
Other Product Names / Synonyms: (S)-3,3'-Dibromo-2,2'-dihydroxy-1,1'-binaphthyl
Formula:
C₂₀H₁₂Br₂O₂ |
MDL Number:
MFCD03093629 CAS Number: 119707-74-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...