(R)-(+)-3,3'-Dibromo-1,1'-bi-2-naphthol ≥98.0%

Supplier: TCI America

D2810-5G D2810-1G
TCD2810-5GEA 1862.82 CAD
TCD2810-5G TCD2810-1G
(R)-(+)-3,3'-Dibromo-1,1'-bi-2-naphthol ≥98.0%
(R)-(+)-3,3'-Dibromo-1,1'-bi-2-naphthol

CAS Number: 111795-43-8
MDL Number: MFCD03093629
Molecular Formula: C20H12Br2O2
Molecular Weight: 444.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Specific rotation [a]20/D: 90 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Asymmetric Synthesis
Other Product Names / Synonyms: (R)-3,3'-Dibromo-2,2'-dihydroxy-1,1'-binaphthyl

Formula: C₂₀H₁₂Br₂O₂
MDL Number: MFCD03093629
CAS Number: 111795-43-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1e71ea9f9fcac35f3da4b6db3b61b042d98ce2d&vid=d1e71ea9f9fcac35f3da4b6db3b61b042d98ce2d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR