trans-1,3-Diphenyl-2-propen-1-ol ≥97.0%

Supplier: TCI America

D2808-25G D2808-5G
TCD2808-25GEA 660.44 CAD
TCD2808-25G TCD2808-5G
trans-1,3-Diphenyl-2-propen-1-ol ≥97.0%
trans-1,3-Diphenyl-2-propen-1-ol

CAS Number: 62668-02-4
MDL Number: MFCD00239426
Molecular Formula: C15H14O
Molecular Weight: 210.28
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 57
Storage Temperature: 0-10°C

Formula: C₁₅H₁₄O
Melting Pt: 57 °C
MDL Number: MFCD00239426
CAS Number: 62668-02-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2cb931a6b7d104819182ac21396ea571938a16b7&vid=2cb931a6b7d104819182ac21396ea571938a16b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR