1,4-Dibromo-2,5-diethylbenzene ≥98.0%

Supplier: TCI America

D2807-1G D2807-5G
TCD2807-1GEA 60.04 CAD
TCD2807-1G TCD2807-5G
1,4-Dibromo-2,5-diethylbenzene ≥98.0%
1,4-Dibromo-2,5-diethylbenzene

CAS Number: 40787-48-2
MDL Number: MFCD04038416
Molecular Formula: C10H12Br2
Molecular Weight: 292.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 40
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2,5-Diethyl-1,4-dibromobenzene

Formula: C₁₀H₁₂Br₂
MW: 292.0103 g/mol
Melting Pt: 40 °C
MDL Number: MFCD04038416
CAS Number: 40787-48-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2130b8bf66a9cad24bacdc7126507bf69b188640&vid=2130b8bf66a9cad24bacdc7126507bf69b188640&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR