4-Diethylamino-2-butyn-1-ol ≥95.0%

Supplier: TCI America

D2805-5ML D2805-25ML
TCD2805-5MLEA 230.68 CAD
TCD2805-5ML TCD2805-25ML
4-Diethylamino-2-butyn-1-ol ≥95.0%
4-Diethylamino-2-butyn-1-ol

CAS Number: 10575-25-4
MDL Number: MFCD00671356
Molecular Formula: C8H15NO
Molecular Weight: 141.21
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 100
Specific Gravity (20/20): 0.95

Formula: C₈H₁₅NO
MW: 141.21 g/mol
Density: 0.95 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00671356
CAS Number: 10575-25-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89bbac6b07f0791f33a66d08a82e6886a06dea09&vid=89bbac6b07f0791f33a66d08a82e6886a06dea09&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR