trans-4-Acetamidocyclohexanol ≥98.0%
Supplier: TCI America
A1162-25G
A1162-5G
TCA1162-025GEA
243.32
CAD
TCA1162-025G
TCA1162-5G
trans-4-Acetamidocyclohexanol ≥98.0%
trans-4-Acetamidocyclohexanol
CAS Number: 27489-60-7
MDL Number: MFCD00051422
Molecular Formula: C8H15NO2
Molecular Weight: 157.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 170
Formula:
C₈H₁₅NO₂ Melting Pt: 170 °C |
MDL Number:
MFCD00051422 CAS Number: 27489-60-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...