(R)-3,3'-Dibromo-2,2'-dimethoxy-1,1'-binaphthyl ≥98.0%
Supplier: TCI America
D2801-200MG
D2801-1G
TCD2801-200MGEA
280.92
CAD
TCD2801-200MG
TCD2801-1G
(R)-3,3'-Dibromo-2,2'-dimethoxy-1,1'-binaphthyl ≥98.0%
(R)-3,3'-Dibromo-2,2'-dimethoxy-1,1'-binaphthyl
CAS Number: 75714-59-9
MDL Number: MFCD04038415
Molecular Formula: C22H16Br2O2
Molecular Weight: 472.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 185
Specific rotation [a]20/D: 12 deg (C=1, CHCl3)
Formula:
C₂₂H₁₆Br₂O₂ Melting Pt: 185 °C |
MDL Number:
MFCD04038415 CAS Number: 75714-59-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...