6,6-Diphenylfulvene ≥98.0%

Supplier: TCI America

Synonyms: 5-(Diphenylmethylene)-1,3-cyclopentadiene

D2787-1G D2787-5G
TCD2787-1GEA 77.74 CAD
TCD2787-1G TCD2787-5G
6,6-Diphenylfulvene ≥98.0%
6,6-Diphenylfulvene

CAS Number: 2175-90-8
MDL Number: MFCD00001419
Molecular Formula: C18H14
Molecular Weight: 230.31
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 81


Other Product Names / Synonyms: 5-(Diphenylmethylene)-1,3-cyclopentadiene

Formula: C₁₈H₁₄
Melting Pt: 81 °C
MDL Number: MFCD00001419
CAS Number: 2175-90-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b81378d62ec2d9c018ea0bcd869b9a3626168c6&vid=4b81378d62ec2d9c018ea0bcd869b9a3626168c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR