6-(Dimethylamino)fulvene ≥98.0%
Supplier: TCI America
D2785-1G
D2785-5G
TCD2785-1GEA
243.32
CAD
TCD2785-1G
TCD2785-5G
6-(Dimethylamino)fulvene ≥98.0%
6-(Dimethylamino)fulvene
CAS Number: 696-68-4
MDL Number: MFCD00013275
Molecular Formula: C8H11N
Molecular Weight: 121.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 67
Storage Temperature: <0°C
Other Product Names / Synonyms: 5-(Dimethylaminomethylene)-1,3-cyclopentadiene
Formula:
C₈H₁₁N Melting Pt: 67 °C |
MDL Number:
MFCD00013275 CAS Number: 696-68-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...