1,4-Dipropylbenzene ≥97.0%

Supplier: TCI America

D2780-25ML D2780-5ML
TCD2780-25MLEA 836.45 CAD
TCD2780-25ML TCD2780-5ML
1,4-Dipropylbenzene ≥97.0%
1,4-Dipropylbenzene

CAS Number: 4815-57-0
MDL Number: MFCD03844789
Molecular Formula: C12H18
Molecular Weight: 162.28
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 221
Specific Gravity (20/20): 0.86

Formula: C₁₂H₁₈
Boiling Pt: 221 °C
Density: 0.86 (20 °C)
MDL Number: MFCD03844789
CAS Number: 4815-57-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d640a14dc940de5eb732e77d10125873f4f4c47&vid=6d640a14dc940de5eb732e77d10125873f4f4c47&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR