(±)-6,6'-Dibromo-1,1'-bi-2-naphthol ≥98.0%

Supplier: TCI America

Synonyms: racemic-6,6'-Dibromo-1,1'-bi-2-naphthol, 6,6'-Dibromo[1,1'-binaphthalene]-2,2'-diol

D2779-1G D2779-5G
TCD2779-1GEA 109.02 CAD
TCD2779-1G TCD2779-5G
(±)-6,6'-Dibromo-1,1'-bi-2-naphthol ≥98.0%
(±)-6,6'-Dibromo-1,1'-bi-2-naphthol

CAS Number: 13185-00-7
MDL Number: MFCD00798290
Molecular Formula: C20H12Br2O2
Molecular Weight: 444.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 205


Other Product Names / Synonyms: (+/-)-6,6'-Dibromo-2,2'-dihydroxy-1,1'-binaphthyl

Formula: C₂₀H₁₂Br₂O₂
Melting Pt: 205 °C
MDL Number: MFCD00798290
CAS Number: 13185-00-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5657b3df0670af8e3994d01114282b2eeaf7e2c0&vid=5657b3df0670af8e3994d01114282b2eeaf7e2c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR