2,2-Diisobutyl-1,3-propanediol ≥98.0%
Supplier: TCI America
D2777-5G
D2777-25G
TCD2777-5GEA
277.76
CAD
TCD2777-5G
TCD2777-25G
2,2-Diisobutyl-1,3-propanediol ≥98.0%
2,2-Diisobutyl-1,3-propanediol
CAS Number: 10547-96-3
MDL Number: MFCD00986394
Molecular Formula: C11H24O2
Molecular Weight: 188.31
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 160
Melting point (°C): 77
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 2,2-Dihydroxy-2,2-diisobutylpropane
Formula:
C₁₁H₂₄O₂ Boiling Pt: 160 °C Melting Pt: 77 °C |
MDL Number:
MFCD00986394 CAS Number: 10547-96-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...