2-Amino-3-chloro-5-(trifluoromethyl)pyridine ≥98.0%

Supplier: TCI America

A1157-5G A1157-25G
TCA1157-005GEA 62.88 CAD
TCA1157-005G TCA1157-025G
2-Amino-3-chloro-5-(trifluoromethyl)pyridine ≥98.0%
2-Amino-3-chloro-5-(trifluoromethyl)pyridine

CAS Number: 79456-26-1
MDL Number: MFCD00042154
Molecular Formula: C6H4ClF3N2
Molecular Weight: 196.56
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 96

Formula: C₆H₄ClF₃N₂
Melting Pt: 96 °C
MDL Number: MFCD00042154
CAS Number: 79456-26-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8a24317a9d17b7ecba390b2933ceac71390ae11&vid=d8a24317a9d17b7ecba390b2933ceac71390ae11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR