Dibenzo[b,e][1,4]dioxine ≥99.0%

Supplier: TCI America

D2776-1G D2776-5G
TCD2776-1GEA 292.3 CAD
TCD2776-1G TCD2776-5G
Dibenzo[b,e][1,4]dioxine ≥99.0%
Dibenzo[b,e][1,4]dioxine

CAS Number: 262-12-4
MDL Number: MFCD00022282
Molecular Formula: C12H8O2
Molecular Weight: 184.19
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 120


Other Product Names / Synonyms: Dibenzo-p-dioxine

Formula: C₁₂H₈O₂
Melting Pt: 120 °C
MDL Number: MFCD00022282
CAS Number: 262-12-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66f6bde19678fb4e4664e83fe70a2f86fdfc692c&vid=66f6bde19678fb4e4664e83fe70a2f86fdfc692c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR