Ethyl-5,6-dichloronicotinate ≥98.0%
Supplier: TCI America
Synonyms:
5,6-Dichloronicotinic Acid Ethyl Ester, 5,6-Dichloropyridine-3-carboxylic Acid Ethyl Ester, Ethyl 5,6-dichloronicotinate, Ethyl 5,6-Dichloropyridine-3-carboxylate
CAS Number: 401566-69-6
MDL Number: MFCD03093627
Molecular Formula: C8H7Cl2NO2
Molecular Weight: 220.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 46
Other Product Names / Synonyms: 5,6-Dichloronicotinic Acid Ethyl Ester, 5,6-Dichloropyridine-3-carboxylic Acid Ethyl Ester, Ethyl 5,6-Dichloropyridine-3-carboxylate
Formula:
C₈H₇Cl₂NO₂ MW: 220.05 g/mol Melting Pt: 46 °C |
MDL Number:
MFCD03093627 CAS Number: 401566-69-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...