2,6-Dihydroxy-4-methylbenzoic acid
Supplier: TCI America
Synonyms:
2,6-Dihydroxy-p-toluic Acid
D2770-25G
D2770-5G
TCD2770-25GEA
341.28
CAD
TCD2770-25G
TCD2770-5G
2,6-Dihydroxy-4-methylbenzoic acid
2,6-Dihydroxy-4-methylbenzoic acid
CAS Number: 480-67-1
MDL Number: MFCD00184686
Molecular Formula: C8H8O4
Molecular Weight: 168.15
Form: Crystal
Color: White
Other Product Names / Synonyms: 2,6-Dihydroxy-p-toluic Acid
Formula:
C₈H₈O₄ MW: 168.15 g/mol |
MDL Number:
MFCD00184686 CAS Number: 480-67-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...