4,4'-Diiodo-3,3'-dimethylbiphenyl ≥98.0%

Supplier: TCI America

Synonyms: 4,4'-Diiodo-3,3'-dimethyl-1,1'-biphenyl

D2759-5G D2759-25G
TCD2759-5GEA 360.24 CAD
TCD2759-5G TCD2759-25G
4,4'-Diiodo-3,3'-dimethylbiphenyl ≥98.0%
4,4'-Diiodo-3,3'-dimethylbiphenyl

CAS Number: 7583-27-9
MDL Number: MFCD04038414
Molecular Formula: C14H12I2
Molecular Weight: 434.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 108

Formula: C₁₄H₁₂I₂
MW: 434.06 g/mol
Melting Pt: 108 °C
MDL Number: MFCD04038414
CAS Number: 7583-27-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=876dc644e751c2d59e5f957d460946b59227eb6b&vid=876dc644e751c2d59e5f957d460946b59227eb6b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR