3-Hydroxytyrosol ≥98.0%

Supplier: TCI America

Synonyms: 2-(3,4-Dihydroxyphenyl)ethanol, Hydroxytyrosol, DOPET, Homoprotocatechuyl alcohol, 2-(3,4-Dihydroxyphenyl)ethyl alcohol, 3,4-Dihydroxyphenethyl Alcohol

D2756-5G D2756-1G
TCD2756-5GEA 1641.3 CAD
TCD2756-5G TCD2756-1G
3-Hydroxytyrosol ≥98.0%
3-Hydroxytyrosol

CAS Number: 10597-60-1
MDL Number: MFCD01320529
Molecular Formula: C8H10O3
Molecular Weight: 154.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid


Other Product Names / Synonyms: 3,4-Dihydroxyphenethyl Alcohol, 2-(3,4-Dihydroxyphenyl)ethanol

Formula: C₈H₁₀O₃
MDL Number: MFCD01320529
CAS Number: 10597-60-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0f86ca2ddd87323d1dd6aa6b261e87dfaf06256&vid=c0f86ca2ddd87323d1dd6aa6b261e87dfaf06256&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR