5,5'-Dibromo-2,2'-bithiophene ≥98.0%
Supplier: TCI America
D2755-1G
D2755-5G
TCD2755-1GEA
75.84
CAD
TCD2755-1G
TCD2755-5G
5,5'-Dibromo-2,2'-bithiophene ≥98.0%
5,5'-Dibromo-2,2'-bithiophene
CAS Number: 4805-22-5
MDL Number: MFCD00219110
Molecular Formula: C8H4Br2S2
Molecular Weight: 324.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 146
Formula:
C₈H₄Br₂S₂ Melting Pt: 146 °C |
MDL Number:
MFCD00219110 CAS Number: 4805-22-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...