3,4-Di-O-acetyl-6-deoxy-L-glucal ≥95.0%
Supplier: TCI America
D2752-1G
D2752-5G
TCD2752-1GEA
75.84
CAD
TCD2752-1G
TCD2752-5G
3,4-Di-O-acetyl-6-deoxy-L-glucal ≥95.0%
3,4-Di-O-acetyl-6-deoxy-L-glucal
CAS Number: 34819-86-8
MDL Number: MFCD00074970
Molecular Formula: C10H14O5
Molecular Weight: 214.22
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.14
Specific rotation [a]20/D: 62 deg (neat)
Product Categories: Bioscience, Glucose, Glycals
Other Product Names / Synonyms: 3,4-Di-O-acetyl-1,5-anhydro-2,6-dideoxy-L-arabino-hex-1-enitol, 3,4-Di-O-acetyl-L-rhamnal
Formula:
C₁₀H₁₄O₅ Density: 1.14 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00074970 CAS Number: 34819-86-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...