4-(Di-p-tolylamino)benzaldehyde ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
4-[Bis(4-methylphenyl)amino]benzaldehyde, 4-(Di-p-tolyl-amino)-benzaldehyde, 4-Formyl-4',4''-dimethyltriphenylamine
CAS Number: 42906-19-4
MDL Number: MFCD03093257
Molecular Formula: C21H19NO
Molecular Weight: 301.39
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 109
Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 4-[Bis(4-methylphenyl)amino]benzaldehyde, 4-Formyl-4',4''-dimethyltriphenylamine
Formula:
C₂₁H₁₉NO MW: 301.39 g/mol Melting Pt: 109 °C |
MDL Number:
MFCD03093257 CAS Number: 42906-19-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...