Methyl-3,4-dihydroxyphenylacetate ≥98.0%
Supplier: TCI America
D2734-5G
D2734-1G
TCD2734-5GEA
347.6
CAD
TCD2734-5G
TCD2734-1G
Methyl-3,4-dihydroxyphenylacetate ≥98.0%
Methyl-3,4-dihydroxyphenylacetate
CAS Number: 25379-88-8
MDL Number: MFCD00583539
Molecular Formula: C9H10O4
Molecular Weight: 182.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 55
Other Product Names / Synonyms: Catechol-4-acetic Acid Methyl Ester, 3,4-Dihydroxyphenylacetic Acid Methyl Ester, Homoprotocatechuic Acid Methyl Ester, Pyrocatechol-4-acetic Acid Methyl Ester
Formula:
C₉H₁₀O₄ Melting Pt: 55 °C |
MDL Number:
MFCD00583539 CAS Number: 25379-88-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...