N-(2-Aminoethyl)glycine ≥97.0% (by titrimetric analysis)

Supplier: TCI America

A1153-100MG A1153-1G
TCA1153-100MGEA 212.13 CAD
TCA1153-100MG TCA1153-001G
N-(2-Aminoethyl)glycine ≥97.0% (by titrimetric analysis)
N-(2-Aminoethyl)glycine

CAS Number: 24123-14-6
MDL Number: MFCD00144824
Molecular Formula: C4H10N2O2
Molecular Weight: 118.14
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 143


Other Product Names / Synonyms: Ethylenediamine-N-monoacetic Acid

Formula: C₄H₁₀N₂O₂
Melting Pt: 143 °C
MDL Number: MFCD00144824
CAS Number: 24123-14-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=668ee6b85789c3f994b28b18ccae7b3131bafab2&vid=668ee6b85789c3f994b28b18ccae7b3131bafab2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR