(S)-(+)-6,6'-Dibromo-1,1'-bi-2-naphthol ≥98.0%

Supplier: TCI America

Synonyms: (S)-6,6'-Dibromo-[1,1'-binaphthalene]-2,2'-diol

D2730-1G D2730-5G
TCD2730-001GEA 222.78 CAD
TCD2730-001G TCD2730-5G
(S)-(+)-6,6'-Dibromo-1,1'-bi-2-naphthol ≥98.0%
(S)-(+)-6,6'-Dibromo-1,1'-bi-2-naphthol

CAS Number: 80655-81-8
MDL Number: MFCD00798290
Molecular Formula: C20H12Br2O2
Molecular Weight: 444.12
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 45 deg (C=2, THF)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis
Other Product Names / Synonyms: (S)-(+)-6,6'-Dibromo-2,2'-dihydroxy-1,1'-binaphthyl

Formula: C₂₀H₁₂Br₂O₂
MDL Number: MFCD00798290
CAS Number: 80655-81-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d041e26ebf60e2fab1fa94328ce7ceab3f6cb6f3&vid=d041e26ebf60e2fab1fa94328ce7ceab3f6cb6f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR