3,4-Dimethoxythiophene ≥98.0%

Supplier: TCI America

D2728-5G D2728-1G
TCD2728-5GEA 218.04 CAD
TCD2728-5G TCD2728-001G
3,4-Dimethoxythiophene ≥98.0%
3,4-Dimethoxythiophene

CAS Number: 51792-34-8
MDL Number: MFCD01096546
Molecular Formula: C6H8O2S
Molecular Weight: 144.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 107

Formula: C6H8O2S
Flash Pt: 107 °C
MDL Number: MFCD01096546
CAS Number: 51792-34-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62cb1d6833186965cadee2115395c7b74b50be8c&vid=62cb1d6833186965cadee2115395c7b74b50be8c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR