1,4-Diamino-2,3-dichloro-9,10-anthraquinone ≥93.0% (by HPLC)

Supplier: TCI America

Synonyms: 1,4-Diamino-2,3-dichloroanthraquinone

D2725-5G D2725-25G
TCD2725-5GEA 30.02 CAD
TCD2725-5G TCD2725-25G
1,4-Diamino-2,3-dichloro-9,10-anthraquinone ≥93.0% (by HPLC)
1,4-Diamino-2,3-dichloro-9,10-anthraquinone

CAS Number: 81-42-5
MDL Number: MFCD00035693
Molecular Formula: C14H8Cl2N2O2
Molecular Weight: 307.13
Purity/Analysis Method: >93.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: Disperse Violet 28

Formula: C₁₄H₈Cl₂N₂O₂
MW: 307.1315 g/mol
MDL Number: MFCD00035693
CAS Number: 81-42-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f9fa9fea40413b5cbd1891706361dc7a3d7c5ce&vid=7f9fa9fea40413b5cbd1891706361dc7a3d7c5ce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR